3-Chloro-6-hydroxypicolinic acid 95%

Supplier: Ambeed
Warning

A164837-100MG A164837-1G A164837-250MG
76824-238EA 54 USD
76824-238 76824-240 76824-242
3-Chloro-6-hydroxypicolinic acid 95%
3-Chloro-6-hydroxypicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MDL Number: MFCD02234757
CAS Number: 1263280-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR