2-tert-Butyl-4-ethylphenol 98%
Supplier: Ambeed
|
Warning
|
A164737-25G
A164737-5G
A164737-500G
A164737-100G
76824-226EA
15
USD
76824-226
76824-228
77550-050
77885-304
2-tert-Butyl-4-ethylphenol 98%
2-tert-Butyl-4-ethylphenol
Precaución: for Research Use Only
Formula:
C₁₂H₁₈O |
MDL Number:
MFCD00069413 CAS Number: 96-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...