2-(3-Chloro-5-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A164566-100MG A164566-1G A164566-250MG
76824-220EA 37.14 USD
76824-220 76824-222 76824-224
2-(3-Chloro-5-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Chloro-5-(cyclopropylmethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₂BClO₃
MW: 308.61 g/mol
MDL Number: MFCD12546580
CAS Number: 1218789-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR