1,4-Dioxaspiro[4.5]decane-8-methanol 95%

Supplier: Ambeed
Warning

A164298-10G A164298-1G A164298-25G A164298-5G A164298-100MG A164298-250MG
76824-204EA 107 USD
76824-204 76824-206 76824-208 76824-210 77885-300 77885-301
1,4-Dioxaspiro[4.5]decane-8-methanol 95%
1,4-Dioxaspiro[4.5]decane-8-methanol

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
MDL Number: MFCD07779441
CAS Number: 17159-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR