(S)-5-Methylmorpholin-3-one 98%

Supplier: Ambeed
Warning

A163474-100MG A163474-1G A163474-250MG A163474-5G
76824-174EA 17 USD
76824-174 76824-176 76824-178 76824-180
(S)-5-Methylmorpholin-3-one 98%
(S)-5-Methylmorpholin-3-one

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MDL Number: MFCD12547742
CAS Number: 119844-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR