Butyl nicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Nicotinic acid butyl ester, Butyl 3-pyridinecarboxylate, n-Butyl nicotinate

A163391-100G A163391-25G A163391-500G A163391-5G
76824-166EA 24 USD
76824-166 76824-168 76824-170 76824-172
Butyl nicotinate 97%
Butyl nicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00006390
CAS Number: 6938-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR