Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A163275-100MG A163275-1G A163275-250MG A163275-5G
76824-150EA 61.54 USD
76824-150 76824-152 76824-154 76824-156
Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate 97%
Ethyl 4,5,6,7-tetrahydro-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
MDL Number: MFCD21364708
CAS Number: 792848-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR