Methyl 5-chloro-2-oxo-2,3-dihydropyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A163267-100MG A163267-250MG
76824-146EA 29.71 USD
76824-146 76824-148
Methyl 5-chloro-2-oxo-2,3-dihydropyrimidine-4-carboxylate 95%
Methyl 5-chloro-2-oxo-2,3-dihydropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₃
MW: 188.57 g/mol
CAS Number: 70662-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR