5,7-Dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-methoxy-4H-chromen-4-one 99+%

Supplier: Ambeed
Warning

A163209-10MG A163209-1MG A163209-25MG A163209-50MG A163209-5MG
76824-136EA 135.8 USD
76824-136 76824-138 76824-140 76824-142 76824-144
5,7-Dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-methoxy-4H-chromen-4-one 99+%
5,7-Dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-6-methoxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₇H₁₄O₇
MW: 330.29 g/mol
MDL Number: MFCD01081948
CAS Number: 18085-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR