(R)-b-Amino-4-fluoro-benzeneethanol 97%

Supplier: Ambeed
Warning

A162457-100MG A162457-1G A162457-250MG A162457-5G
76824-118EA 20 USD
76824-118 76824-120 76824-122 76824-124
(R)-b-Amino-4-fluoro-benzeneethanol 97%
(R)-b-Amino-4-fluoro-benzeneethanol

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
MDL Number: MFCD09253715
CAS Number: 174770-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR