Methyl 3-fluoro-5-hydroxy-4-methylbenzoate 95+%

Supplier: Ambeed
Warning

A161790-1G A161790-250MG
76824-106EA 82.76 USD
76824-106 76824-108
Methyl 3-fluoro-5-hydroxy-4-methylbenzoate 95+%
Methyl 3-fluoro-5-hydroxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.16 g/mol
MDL Number: MFCD11042808
CAS Number: 1215031-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR