2-Amino-N-methylnicotinamide 95%

Supplier: Ambeed
Warning

A161667-1G A161667-250MG A161667-100MG
76824-102EA 52 USD
76824-102 76824-104 77885-296
2-Amino-N-methylnicotinamide 95%
2-Amino-N-methylnicotinamide

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MDL Number: MFCD11520028
CAS Number: 870997-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR