Thiophene-2-carbaldehyde 98% stabilized
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde
Stabilization:
Stabilised with hydroquinone (HQ)
A161612-1000G
A161612-100G
A161612-25G
A161612-500G
76824-094EA
51.99
USD
76824-094
76824-096
76824-098
76824-100
Thiophene-2-carbaldehyde 98% stabilized
Thiophene-2-carbaldehyde
Precaución: for Research Use Only
Formula:
C₅H₄OS MW: 112.15 g/mol |
MDL Number:
MFCD00005429 CAS Number: 98-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...