Benzyl 3,3-difluoro-4-oxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A161260-100MG A161260-250MG
76824-084EA 77.45 USD
76824-084 76824-086
Benzyl 3,3-difluoro-4-oxopiperidine-1-carboxylate 95%
Benzyl 3,3-difluoro-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃F₂NO₃
MW: 269.244 g/mol
MDL Number: MFCD16250753
CAS Number: 1283720-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR