Imidazo[1,2-a]pyridin-5-ylmethanol 95%

Supplier: Ambeed
Warning

A159752-5G A159752-250MG A159752-1G
76824-066EA 511.36 USD
76824-066 77592-944 77683-331
Imidazo[1,2-a]pyridin-5-ylmethanol 95%
Imidazo[1,2-a]pyridin-5-ylmethanol

Precaución: For research use only.

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD13183181
CAS Number: 167884-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR