7-Benzyloxy-6-methoxy-3H-quinazolin-4-one 97%

Supplier: Ambeed
Warning

Synonyms: 7-Benzyloxy-4-hydroxy-6-methoxyquinazoline, 7-Benzyloxy-6-methoxy-4-quinazolinol, 7-(Benzyloxy)-6-methoxyquinazolin-4(3H)-one

A159675-100G A159675-10G A159675-1G A159675-25G A159675-5G
76824-056EA 181.42 USD
76824-056 76824-058 76824-060 76824-062 76824-064
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one 97%
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂O₃
MW: 282.30 g/mol
MDL Number: MFCD04115119
CAS Number: 179688-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR