6-Methoxy-1-benzofuran 96%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxybenzofuran

A158940-250MG A158940-5G A158940-100MG
76824-050EA 55.17 USD
76824-050 76824-052 77683-325
6-Methoxy-1-benzofuran 96%
6-Methoxy-1-benzofuran

Precaución: For research use only.

Formula: C₉H₈O₂
MW: 148.159 g/mol
MDL Number: MFCD11656393
CAS Number: 50551-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR