3-Bromo-1H-indazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A158816-100MG A158816-1G A158816-250MG A158816-5G
76824-044EA 7.43 USD
76824-044 76824-046 76824-048 77611-528
3-Bromo-1H-indazole-5-carboxylic acid 95%
3-Bromo-1H-indazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.04 g/mol
MDL Number: MFCD07781587
CAS Number: 885521-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR