(3S)-3-(Methoxymethyl)morpholine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(Methoxymethyl)morpholine hydrochloride

A158488-100MG A158488-1G A158488-250MG A158488-5G
76824-036EA 24.41 USD
76824-036 76824-038 76824-040 76824-042
(3S)-3-(Methoxymethyl)morpholine hydrochloride 97%
(3S)-3-(Methoxymethyl)morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂·ClH
MW: 167.634 g/mol
MDL Number: MFCD09608107
CAS Number: 218594-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR