(R)-2-Acetamido-2-(2-fluorophenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A158304-250MG A158304-1G
76824-026EA 23 USD
76824-026 77885-281
(R)-2-Acetamido-2-(2-fluorophenyl)propanoic acid 95%
(R)-2-Acetamido-2-(2-fluorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO₃
MW: 225.22 g/mol
MDL Number: MFCD21364763
CAS Number: 267401-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR