(4-Butoxy-3-chlorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Butoxy-3-chlorophenylboronic acid

A158221-1G A158221-250MG A158221-25G A158221-5G
76824-018EA 40 USD
76824-018 76824-020 76824-022 76824-024
(4-Butoxy-3-chlorophenyl)boronic acid 95%
(4-Butoxy-3-chlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClO₃
MDL Number: MFCD04974064
CAS Number: 480438-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR