2-(2-Bromophenyl)propan-2-amine 95%

Supplier: Ambeed
Warning

A157559-100MG A157559-250MG
76823-996EA 100.79 USD
76823-996 76823-998
2-(2-Bromophenyl)propan-2-amine 95%
2-(2-Bromophenyl)propan-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.10 g/mol
MDL Number: MFCD11109721
CAS Number: 173026-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR