1,5-Dimethyl-2-(p-tolyl)-1H-pyrazol-3(2H)-one 97%

Supplier: Ambeed
Warning

A156801-1G A156801-250MG
76823-980EA 24 USD
76823-980 76823-982
1,5-Dimethyl-2-(p-tolyl)-1H-pyrazol-3(2H)-one 97%
1,5-Dimethyl-2-(p-tolyl)-1H-pyrazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O
MDL Number: MFCD00003140
CAS Number: 56430-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR