Methyl 6-chloro-4-methoxypicolinate 98%

Supplier: Ambeed
Warning

A155635-100MG A155635-250MG A155635-5G
76823-952EA 21.22 USD
76823-952 76823-954 76823-956
Methyl 6-chloro-4-methoxypicolinate 98%
Methyl 6-chloro-4-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.61 g/mol
MDL Number: MFCD12756111
CAS Number: 204378-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR