5-Bromo-6-iodopyridin-2-amine 95%

Supplier: Ambeed
Warning

A154530-250MG A154530-1G
76823-936EA 70 USD
76823-936 77549-960
5-Bromo-6-iodopyridin-2-amine 95%
5-Bromo-6-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
MDL Number: MFCD14708215
CAS Number: 1223748-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR