6-Chloroisobenzofuran-1(3H)-one 95%

Supplier: Ambeed
Warning

A154461-100MG A154461-1G A154461-250MG A154461-5G
76823-928EA 15 USD
76823-928 76823-930 76823-932 76823-934
6-Chloroisobenzofuran-1(3H)-one 95%
6-Chloroisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅ClO₂
MDL Number: MFCD04108605
CAS Number: 19641-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR