Methyl 2-(5-bromo-2-chlorophenyl)acetate 98%

Supplier: Ambeed
Warning

A153329-1G A153329-250MG A153329-5G A153329-25G
76823-894EA 17 USD
76823-894 76823-896 77611-450 77885-257
Methyl 2-(5-bromo-2-chlorophenyl)acetate 98%
Methyl 2-(5-bromo-2-chlorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MDL Number: MFCD16038439
CAS Number: 203314-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR