2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethylphenylboronic acid Pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-o-xylene

A153313-1G A153313-250MG A153313-10G
76823-890EA 23.34 USD
76823-890 76823-892 77683-287
2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₁₄H₂₁BO₂
MW: 232.13 g/mol
MDL Number: MFCD12405519
CAS Number: 401797-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR