(S)-tert-Butyl 6-amino-2-((tert-butoxycarbonyl)amino)hexanoate hydrochloride 97%

Supplier: Ambeed
Warning

A152323-1G A152323-250MG
76823-862EA 83 USD
76823-862 77885-249
(S)-tert-Butyl 6-amino-2-((tert-butoxycarbonyl)amino)hexanoate hydrochloride 97%
(S)-tert-Butyl 6-amino-2-((tert-butoxycarbonyl)amino)hexanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₃₁ClN₂O₄
MDL Number: MFCD08275791
CAS Number: 7750-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR