6-Chloro-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A151932-100MG A151932-250MG A151932-1G
76823-858EA 46.68 USD
76823-858 76823-860 77683-280
6-Chloro-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde 95%
6-Chloro-1H-pyrrolo[3,2-c]pyridine-3-carbaldehyde

Precaución: For research use only.

Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
MDL Number: MFCD09749983
CAS Number: 1000341-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR