1-Bromo-3-iodo-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A151633-1G A151633-250MG A151633-25G
76823-850EA 39 USD
76823-850 76823-852 77885-247
1-Bromo-3-iodo-5-nitrobenzene 97%
1-Bromo-3-iodo-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrINO₂
MDL Number: MFCD13194576
CAS Number: 861601-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR