5-Methyl-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 97%

Supplier: Ambeed
Warning

A151472-100MG A151472-250MG
76823-838EA 9 USD
76823-838 76823-840
5-Methyl-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 97%
5-Methyl-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₇N₃O
MDL Number: MFCD12923894
CAS Number: 919278-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR