(1R,2S)-Ethyl 1-((tert-butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylate 97%

Supplier: Ambeed
Warning

A150387-10G A150387-1G
76823-812EA 245.07 USD
76823-812 76823-814
(1R,2S)-Ethyl 1-((tert-butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylate 97%
(1R,2S)-Ethyl 1-((tert-butoxycarbonyl)amino)-2-vinylcyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₄
MW: 255.31 g/mol
MDL Number: MFCD06795881
CAS Number: 259217-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR