4-Methoxy-7-morpholinobenzo[d]thiazol-2-amine 95%

Supplier: Ambeed
Warning

A149807-250MG A149807-100MG A149807-1G
76823-804EA 79 USD
76823-804 77885-237 77885-238
4-Methoxy-7-morpholinobenzo[d]thiazol-2-amine 95%
4-Methoxy-7-morpholinobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₂S
MDL Number: MFCD18642707
CAS Number: 383865-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR