6-Chloro-2,3-dihydroquinolin-4(1H)-one 97%

Supplier: Ambeed
Warning

A149432-1G A149432-5G
76823-782EA 127.31 USD
76823-782 76823-784
6-Chloro-2,3-dihydroquinolin-4(1H)-one 97%
6-Chloro-2,3-dihydroquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MDL Number: MFCD09834922
CAS Number: 21617-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR