4-Bromo-3-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-3-methoxyphenyl)boronic acid

A148832-100MG A148832-1G A148832-250MG A148832-5G
76823-760EA 33.95 USD
76823-760 76823-762 76823-764 76823-766
4-Bromo-3-methoxyphenylboronic acid 98%
4-Bromo-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
MW: 230.8500 g/mol
MDL Number: MFCD16036103
CAS Number: 1256345-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR