Cyclopropylboronic acid MIDA ester 97%

Supplier: Ambeed
Warning

A148654-1G A148654-250MG A148654-5G
76823-754EA 49.87 USD
76823-754 76823-756 76823-758
Cyclopropylboronic acid MIDA ester 97%
Cyclopropylboronic acid MIDA ester

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₄
MW: 196.9962 g/mol
MDL Number: MFCD11215247
CAS Number: 1104637-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR