(S)-2-Aminobut-3-enoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Vinyl-Gly-OH·HCl

A147919-100MG A147919-250MG A147919-1G
76823-724EA 199.45 USD
76823-724 76823-726 77549-818
(S)-2-Aminobut-3-enoic acid hydrochloride 97%
(S)-2-Aminobut-3-enoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈ClNO₂
MW: 137.56 g/mol
MDL Number: MFCD01863784
CAS Number: 75266-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR