3-Bromo-8-chloroquinoline 95%

Supplier: Ambeed
Warning

A147537-100MG A147537-250MG
76823-702EA 111.4 USD
76823-702 76823-704
3-Bromo-8-chloroquinoline 95%
3-Bromo-8-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MW: 242.5 g/mol
MDL Number: MFCD28129771
CAS Number: 205111-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR