3'-Chloropropiophenone 98%

Supplier: Ambeed
Warning

A147259-1000G A147259-100G A147259-10G A147259-25G A147259-5G
76823-678EA 79 USD
76823-678 76823-680 76823-682 76823-684 76823-686
3'-Chloropropiophenone 98%
3'-Chloropropiophenone

Precaución: for Research Use Only

Formula: C₉H₉ClO
MDL Number: MFCD00009925
CAS Number: 34841-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR