(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-3-(3-methoxyphenyl)propanoic acid

A146711-5G A146711-250MG A146711-1G
76823-672EA 333.13 USD
76823-672 77611-352 77683-252
(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 98+%
(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid

Precaución: For research use only.

Formula: C₁₅H₂₁NO₅
MW: 295.34 g/mol
MDL Number: MFCD03427937
CAS Number: 500788-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR