2,5-Dibromo-4-methylpyrimidine 97%

Supplier: Ambeed
Warning

A146635-100MG A146635-10G A146635-250MG A146635-5G
76823-664EA 13 USD
76823-664 76823-666 76823-668 76823-670
2,5-Dibromo-4-methylpyrimidine 97%
2,5-Dibromo-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MDL Number: MFCD13193658
CAS Number: 171408-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR