Tetrahydro-2H-pyran-4-yl-4-methylbenzenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Tetrahydro-2H-pyran-4-yl 4-methylbenzenesulphonate

A146416-100MG A146416-1G A146416-250MG
76823-658EA 29.71 USD
76823-658 76823-660 76823-662
Tetrahydro-2H-pyran-4-yl-4-methylbenzenesulfonate 98%
Tetrahydro-2H-pyran-4-yl-4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄S
MW: 256.32 g/mol
MDL Number: MFCD16620977
CAS Number: 97986-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR