Ethyl 2,2-difluoro-2-(2-methoxyphenyl)acetate 95%

Supplier: Ambeed
Warning

A146404-100MG A146404-5G
76823-654EA 31.83 USD
76823-654 76823-656
Ethyl 2,2-difluoro-2-(2-methoxyphenyl)acetate 95%
Ethyl 2,2-difluoro-2-(2-methoxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.21 g/mol
MDL Number: MFCD12026027
CAS Number: 1150164-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR