2,6-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A144667-100MG A144667-1G A144667-250MG
76823-600EA 8 USD
76823-600 76823-602 76823-604
2,6-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₂₅H₂₃N₃O₂
MDL Number: MFCD23704921
CAS Number: 151670-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR