4-Chloropiperidine hydrochloride 97%

Supplier: Ambeed
Warning

A144553-10G A144553-1G
76823-592EA 37 USD
76823-592 77885-211
4-Chloropiperidine hydrochloride 97%
4-Chloropiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁Cl₂N
MW: 156.0500 g/mol
MDL Number: MFCD03411587
CAS Number: 5382-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR