Methyl 4-hydroxy-2,6-dimethylbenzoate 95%
Supplier: Ambeed
|
Warning
|
A144317-100MG
A144317-250MG
A144317-5G
76823-578EA
30
USD
76823-578
76823-580
77885-209
Methyl 4-hydroxy-2,6-dimethylbenzoate 95%
Methyl 4-hydroxy-2,6-dimethylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₃ |
MDL Number:
MFCD13191754 CAS Number: 83194-70-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...