2-Propylvaleric acid 98%

Supplier: Ambeed
Danger

A143794-10G A143794-25G A143794-100G A143794-1000G A143794-500G
76823-564EA 6 USD
76823-564 76823-566 77316-614 77592-700 77611-320 77885-207
2-Propylvaleric acid 98%
2-Propylvaleric acid

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD00002672
CAS Number: 99-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR