5-(4-Bromophenyl)pyrrolidin-2-one 96%

Supplier: Ambeed
Warning

A143198-100MG A143198-250MG A143198-5G
76823-538EA 18.04 USD
76823-538 76823-540 76823-542
5-(4-Bromophenyl)pyrrolidin-2-one 96%
5-(4-Bromophenyl)pyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MW: 240.09 g/mol
MDL Number: MFCD15527558
CAS Number: 207989-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR