Ethyl 4-bromothiophene-2-carboxylate 95%

Supplier: Ambeed
Warning

A143192-100MG A143192-10G A143192-1G A143192-250MG A143192-25G A143192-5G
76823-526EA 5.31 USD
76823-526 76823-528 76823-530 76823-532 76823-534 76823-536
Ethyl 4-bromothiophene-2-carboxylate 95%
Ethyl 4-bromothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
MW: 235.10 g/mol
MDL Number: MFCD06204328
CAS Number: 62224-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR